TAQuest™ FAST qPCR Master Mix with Helixyte™ Green *Low ROX*
Supplier: AAT BIOQUEST INC
17278 17279
77461-586EA 333.43 CAD
77461-586 77461-588
TAQuest™ FAST qPCR Master Mix with Helixyte™ Green *Low ROX*
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
TAQuest™ FAST qPCR Master mix with Helixyte™ green is a ready-to-use 2X solution optimized for qPCR and two-step RT-qPCR.
  • Simple and robust protocol
  • The optimized composition ensures PCR specificity and sensitivity with all sample types such as genomic, plasmid, viral and cDNA templates
The Helixyte green intercalating dye allows rapid DNA detection and analysis without using sequence-specific probes.
Caution: Master mix is stable for 12 months when stored frozen (<−15 °C) with minimized light exposure.
Order Now
SPECIFICATIONS
Application qPCR dye
Environmentally Preferable