Buccutite™ Streptavidin Antibody Conjugation Kit
Supplier: AAT BIOQUEST INC
5511
5509
5510
77462-904EA
809.62
CAD
77462-904
77462-900
77462-902
Buccutite™ Streptavidin Antibody Conjugation Kit
Protein Linking and Labeling Systems
Buccutite™ Streptavidin antibody conjugation kits are optimized for labelling either 25 µg, 100 µg or 1 mg of protein.
- The produced streptavidin-conjugated antibodies may be directly used in WB, ELISA, IHC without further purification
- Can also be used to conjugate other proteins or peptides
- It can be used to generate conjugates of different ratios of streptavidin/antibody
This streptavidin conjugation kit uses a simple and quick process for crosslinking streptavidin to an antibody.
Delivery information: Shipped at RT.
Caution: Research use only.