Anti-IgG Goat Polyclonal Antibody (PE (Phycoerythrin))

Supplier: AAT BIOQUEST INC
50220 50221
77462-748EA 264.11 CAD
77462-748 77462-750
Anti-IgG Goat Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Anti-human secondary antibodies have well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target.

  • Affinity purified
  • Easily reconstituted

Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence).

Caution: For research use only.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol IgG
Environmentally Preferable
Antigen name Immunoglobulin G
Antibody type Secondary
Clonality Polyclonal
Conjugation PE (Phycoerythrin)
Reactivity Human
Host Goat
Isotype IgG (H, L)
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Flow cytometry Yes
Cross adsorption No
Storage temperature Freeze (<–15 °C); minimize light exposure
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR