Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (FITC (Fluorescein Isothiocyanate))

Supplier: AAT BIOQUEST INC
50206 50207
77462-720EA 183.55 CAD
77462-720 77462-722
Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (FITC (Fluorescein Isothiocyanate))
Antibodies
Anti-human secondary antibodies have well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target.

  • Affinity purified
  • Easily reconstituted

Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence).

Caution: For research use only.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR