Fetal Bovine Serum, Premium, New Zealand Origin (Heat Inactivated and Gamma Irradiated)

Supplier: Corning
35-108-CV 35-108-CV
76332-598CS 2195.25 CAD
76332-598 CA76332-598
Fetal Bovine Serum, Premium, New Zealand Origin (Heat Inactivated and Gamma Irradiated)
Sera (Serum)
Species: Bovine, bos taurus
Type: Fetal
Treatment: Heat inactivated and gamma irradiated
Format: Liquid
Shelf life: 60 months

  • Collected at government registered facilities in New Zealand
  • Exported from the country of origin accompanied with a veterinarian health certificate and imported under a USDA-APHIS issued import permit
  • True pooled and is triple 0.1 µm sterile filtered that passes sterility, mycoplasma, and 9CFR viral testing requirements
  • Validated process used to heat inactivate serum at 56 °C (±2 °C) for 30 minutes via bulk inactivation of entire lot in the pooling tank or individual bottles in a water bath
  • Validated process used to gamma irradiate serum within one of two separate dose ranges: 25 to 40 kGy and 30 to 45 kGy
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR