Fetal Bovine Serum, Premium, New Zealand Origin

Supplier: Corning
35-078-CF 35-078-CF
76382-762EA 401.3 CAD
76382-762 CA76382-762
Fetal Bovine Serum, Premium, New Zealand Origin
Sera (Serum)
Species: Bovine, bos taurus
Type: Fetal
Format: Liquid
Shelf life: 60 months

  • Collected at government registered facilities in New Zealand
  • Exported from the country of origin accompanied with a veterinarian health certificate and imported under a USDA-APHIS issued import permit
  • True pooled and triple 0.1 µm sterile filtered that passes sterility, mycoplasma, and 9CFR viral testing requirements
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR