VWR® Vials, Caps, and Kits for PFAS Testing
Supplier: VWR International
89524-176 89524-178 89524-180 89524-182 89524-184 89524-186
89524-176CS 308.54 CAD
89524-176 89524-178 89524-180 89524-182 89524-184 89524-186
VWR® Vials, Caps, and Kits for PFAS Testing
Vials
A key requirement for PFAS testing is using PFAS-free container closure systems (CCS), which consist of a container and a sealing method. Vials and caps are available separately or as kits, providing flexibility for different testing needs.
  • Polypropylene (PP) construction prevents PFAS contamination
  • Individual caps, vials, and kitted configurations available
  • 9 mm screw thread
  • PFAS-free packaging
Government regulations requiring reduction and elimination of PFAS in raw materials are driving the demand for PFAS-free consumables used by analytical labs testing wastewater, drinking water, ground/surface water, air, food, soil, serum, blood, and cosmetics.