XPR Essential Analytical Balances
Supplier: Mettler Toledo
30904667 30904692 30904711 30904716 30904731 30904736 30904741 30923237
CA77581-348EA 19300 CAD
CA77581-348 CA77581-350 CA77581-352 CA77581-354 CA77581-356 CA77581-358 CA77581-360 CA77581-362
XPR Essential Analytical Balances
Balances and Scales
METTLER TOLEDO XPR Essential analytical balances provide reliable results with a high-performance load cell, SmartPan hanging weigh pan, and FACT automatic internal adjustment. One-touch automatic doors simplify dosing, and SmartTrac provides dosing guidance on the terminal. Advanced user management with password protection streamline daily processes. The METTLER TOLEDO XPR Essential Analytical Balances are the successor to the XSR analytical balances.
  • StatusLight: Indicates balance is ready
  • MinWeigh: Warning function for low weights
  • LevelControl: Ensures balance is levelled
  • Compact ionizer (connectable with stand)
  • 7" color touchscreen
  • ErgoClips for dosing
  • SmartTrac: Dosing guidance on terminal
XPR Essential analytical balances feature a digital results notepad, storage of SOPs in method library and LabX connectivity. These models have a compact front-loader design with easy-to-clean draft shields and overload protection for the load cell. The balances are environmentally friendly with carbon neutral operation.
Balance rankings:
XPR Excellence analytical balances
XPR Essential analytical balances
MX Advanced analytical balances
MR Advanced analytical balances
MA Standard analytical balances
LA Standard analytical balances