ReadiPrep™ Protein G-Agarose Resin
Supplier: AAT BIOQUEST INC
55016 55017
77462-896EA 159.49 CAD
77462-896 77462-898
ReadiPrep™ Protein G-Agarose Resin
Protein Purification Protein Purification Resins
ReadiPrep™ Protein G-Agarose Resin is a high-quality affinity resin designed for the isolation and purification of immunoglobulin classes, subclasses, and fragments from mammalian serum and biological fluids.
  • Stable for 12 months
Quality tested in-house.
Caution: For research use only.