Human Recombinant CD33 (from HEK293 cells), RPE (R-Phycoerythrin)

Supplier: Creative Biomart

CD33-459HP-100UG CD33-459HP-10UG CD33-459HP-50UG
77538-922EA 4065.25 CAD
77538-922 77538-924 77538-926
Human Recombinant CD33 (from HEK293 cells), RPE (R-Phycoerythrin)
Proteins and Peptides

Order Now

SPECIFICATIONS

Protein/peptide name CD33
Environmentally Preferable
Protein synonyms CD33 CD33 molecule
Protein/peptide type Recombinant
Species Human
Source HEK293 cells
Conjugation RPE (R-Phycoerythrin)
Protease-free No
Animal-free No
Nuclease-free No
Carrier-free No
Endotoxin-free No
Endotoxin level Low
Endotoxin content <1 EU/µg by LAL method
Grade Ultrapure grade
Storage conditions 4 °C
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR