Autoclave Bags
Supplier: Heathrow Scientific
HS1002A HS1002B HS1002C
77529-450CS 535.74 CAD
77529-450 77529-452 77529-454
Autoclave Bags
Bags Sterilization Pouches
Clear bags imprinted with blue biohazard labeling designed for safe disposal.
  • Constructed of 2.0 mm thick polypropylene
  • Suitable for repeated autoclaving
  • Strong side seams for securely holding items
  • Front write-on panel
  • Clear color finish
Available in 14, 26 and 69 liter sizes.
Delivery information: Sold in cases of 200.