Steri-Perox® wipes leave no residue and are used for both aseptic and non-aseptic wipe downs of filling and packaging machinery, stainless steel surfaces, Lexan, polycarbonate, glass, and any critical surfaces that require cleaning.
- Available with 3 or 6% stabilized hydrogen peroxide solution
- Packaged in ready-to-use sterile containers
- Filtered through a 0.2µm filter and gamma-irradiated to ensure sterility
- Effective against a wide range of microorganisms, including bacteria, viruses, fungi, and spores
- Leaves no harmful residues after application, ensuring a clean and safe environment
- Safe for use on a variety of surfaces in controlled environments
- Provided with a lot-specific Certificate of Analysis and Certificate of Sterility
Steri-Perox® is a 3 or 6% stabilized hydrogen peroxide solution designed for high-level disinfection in critical environments. Filtered and gamma-irradiated to ensure sterility, it is residue-free and effective against a broad spectrum of microorganisms. Targeted for use in pharmaceutical, biotechnology, and healthcare industries, Steri-Perox® is ideal for cleanrooms, aseptic processing areas, and other controlled environments where stringent microbial control is essential.
Steri-Perox® wipes ensure effective microbial control in critical environments in a ready-to-use format that reduces preparation time and risk of contamination. Non-toxic and residue-free, these wipes ensure safety for users and equipment, while supporting compliance with industry standards for cleanroom environments. Each batch is rigorously tested and documented, providing assurance of consistency and effectiveness.
Packaging: Wipes are packaged 20 per pack, 10 packs per case (200 total wipes).