Glossy White 2 mil Polyimide Circuit Board Labels
Supplier: Brady Worldwide
176958 176959 176960 176961 176962 176963 176964 176975 176970 176971 176972 176973 176974
77574-842RL 335.17 CAD
77574-842 77574-878 77574-926 77574-864 77574-920 77574-840 77574-884 77574-868 77574-848 77574-854 77574-910 77574-872 77574-936
Glossy White 2 mil Polyimide Circuit Board Labels
Labels
For PCB and electronic component manufacturing, get labels that are engineered to survive extreme processes like wave soldering and multiple harsh washes.
  • Ultra-durable – adhesive is built for extreme washes and cleaning chemicals
  • Enhanced topcoat is ideal for boards that go through multiple harsh-condition washes
  • Labels do not require reflow or preheatings to ensure printing readability
  • Smear-resistant, glossy white polyimide material (B-797) is thermal transfer printable
  • Service temperature of –70 to +100 °C (–94 to +212 °F)
  • Meets requirements of MIL-STD-202G, Method 215K
  • A ribbon is required to print this material. Use part number M7-R6300 for the M710 / BMP71 label printers
Certifications: UL Approved