Acetyltransferase Dilution Buffers
Supplier: Sino Biological
A21-09-01
A21-09-05
77554-342EA
26.86
CAD
77554-342
77554-344
Acetyltransferase Dilution Buffers
Buffers
Assay Buffers
Solution of buffer used for diluting enzyme histone acetyltransferases (KAT).
- Application: Enzyme assay
For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...