VEE GEE Volumetric Pipettes, Wide Tip
Supplier: Heathrow Scientific
2040-100-3MM 2040-10-3MM 2040-20-3MM 2040-2-2.4MM 2040-25-3MM 2040-5-3MM
76546-046CS 6843.74 CAD
76546-046 76546-048 76546-052 76546-054 76546-056 76546-060
VEE GEE Volumetric Pipettes, Wide Tip
Pipettes Graduated Pipettes
Our wide tip volumetric pipettes are meticulously manufactured for tolerances and accuracy in order to ensure pinpoint precision for all of your liquid handling needs.
  • Made out of high-quality borosilicate glass
  • Wide range of sizes to choose from
  • Reusable
  • Color-coded with permanent enamel markings
  • Calibrated To Deliver
  • Meets ASTM E-969 Class B Specifications and USP Standards
They also have graduation markings and color-coded bands that are permanently enameled to the glass which allows for quick identification of different volume ranges. The wide tipMore Product Information