RPE-Cy5-streptavidin conjugate
Supplier: AAT BIOQUEST INC
16918
16919
77461-510EA
167.89
CAD
77461-510
77461-512
RPE-Cy5-streptavidin conjugate
RPE-Cy5-streptavidin
RPE-Cy5-streptavidin conjugate comprises streptavidin (as the biotin-binding protein) with RPE-Cy5 covalently attached (as the fluorescent label).
- Stable for 12 months
Streptavidin conjugates are widely used together with a biotin conjugate for specific detection of a variety of biological molecules since streptavidin has a very high binding affinity for biotin and its conjugates.
Caution: For Research Use Only