APC maleimide, ReadiUse™, preactivated [Activated Allophycocyanin]
Supplier: AAT BIOQUEST INC
2567
2568
77462-044EA
248.08
CAD
77462-044
77462-046
APC maleimide, ReadiUse™, preactivated [Activated Allophycocyanin]
APC maleimide
ReadiUse™ Preactivated APC Maleimide is prepared by reacting highly purified allophycocyanin (APC) with SMCC.
- Stable for 6 months
Quality tested in-house
Caution: For Research Use Only