MTS reagent 5 mM in aqueous solution, ReadiUse™

Supplier: AAT BIOQUEST INC

15710 15711
77461-490EA 299.38 CAD
77461-490 77461-492
MTS reagent 5 mM in aqueous solution, ReadiUse™
MTS reagent

MTS reagent is one of the most common water-soluble tetrazolium reagents that are widely used to quantify cell viability by absorption spectrophotometry.


  • Stable for 6 months


Quality tested in-house


Caution: For Research Use Only

Formula: C₂₀H₁₇N₅O₆S₂
CAS Number: 138169-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR