Psoralen-A17-Biotin, ReadiLink™
Supplier: AAT BIOQUEST INC
39052 39053
77462-234EA 209.21 CAD
77462-234 77462-236
Psoralen-A17-Biotin, ReadiLink™
Psoralen-A17-Biotin
ReadiLink™ Psoralen-A17-Biotin is a photoactivatable reagent for biotinylating DNA or RNA.
  • Stable for 24 months
Quality tested in-house
Caution: For Research Use Only