Psoralen-PEG3-Biotin
Supplier: AAT BIOQUEST INC
39050 39051
77462-230EA 159.49 CAD
77462-230 77462-232
Psoralen-PEG3-Biotin
Psoralen-PEG3-Biotin
Psoralen-PEG3-Biotin is a photoactivatable reagent for biotinylating DNA or RNA.
  • Stable for 24 months
Quality tested in-house
Caution: For Research Use Only