MycoLight™ Green JJ98
Supplier: AAT BIOQUEST INC
23998
23999
77461-932EA
256.47
CAD
77461-932
77461-934
MycoLight™ Green JJ98
MycoLight™ Green JJ98
MycoLight™ Green JJ98 stain is an excellent green-fluorescent nuclear and chromosome counterstain that is permeant to both prokaryotic and eukaryotic cell membranes.
- Stable for 12 months
Quality tested in-house
Caution: For Research Use Only