iFluor® Ultra 594 succinimidyl ester
Supplier: AAT BIOQUEST INC
71650
71651
71652
77463-056EA
517.01
CAD
77463-056
77463-058
77463-060
iFluor® Ultra 594 succinimidyl ester
iFluor® Ultra 594 succinimidyl ester
iFluor® Ultra 594 is an exceptionally bright red fluorescent dye excitable by 561 nm or 594 nm laser lines, with a peak emission at 601 nm.
- Stable for 12 months
Fluorescent dye-conjugated antibodies provide a tool for identifying proteins in many applications including fluorescent cell imaging, flow cytometry, western blotting, immunohistochemistry and more.
Caution: For Research Use Only