iFluor® 830 succinimidyl ester
Supplier: AAT BIOQUEST INC
1384 71400 71530 71580
77461-468EA 558.7 CAD
77461-468 77463-050 77463-052 77463-054
iFluor® 830 succinimidyl ester
iFluor® 830 succinimidyl ester
iFluor® 830 dye emission is a unique color for spectrum flow cytometry as it is well separated from commonly used far-red fluorophores such as Cy5, Cy7, or allophycocyanin (APC), facilitating multicolor analysis.
  • Stable for 12 months
In vivo fluorescence imaging uses a sensitive camera to detect the fluorescence emission from fluorophores in whole-body living small animals.
Caution: For Research Use Only