iFluor® 780 maleimide
Supplier: AAT BIOQUEST INC
1372
77461-462EA
558.7
CAD
77461-462
iFluor® 780 maleimide
iFluor® 780 maleimide
iFluor® 780 dyes have fluorescence excitation and emission maxima of ~784 nm and ~808 nm respectively.
- Stable for 12 months
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, particularly antibodies.
Caution: For Research Use Only