iFluor® 605 succinimidyl ester
Supplier: AAT BIOQUEST INC
1420
77461-472EA
371.92
CAD
77461-472
iFluor® 605 succinimidyl ester
iFluor® 605 succinimidyl ester
iFluor® 605 succinimidyl ester is the most convenient reactive form of iFluor® 605 dyes that can be readily used for labeling proteins such as antibodies.
- Stable for 12 months
Quality tested in-house
Caution: For Research Use Only