Helixyte™ Green ssDNA reagent
Supplier: AAT BIOQUEST INC
17620 17621
77461-646EA 341.44 CAD
77461-646 77461-648
Helixyte™ Green ssDNA reagent
Helixyte™ Green
Helixyte™ Green ssDNA Quantifying Reagent is an excellent alternative for quantifying ssDNA and oligonucleotides with significantly improved sensitivity and selectivity.
  • Stable for 12 months
Synthetic oligonucleotides are used in several molecular biology techniques, such as DNA sequencing, site-directed mutagenesis, DNA amplification, and in situ hybridization.
Caution: For Research Use Only