FAM-xtra CPG 1000 Å
Supplier: AAT BIOQUEST INC
6043 6044
77462-960EA 331.83 CAD
77462-960 77462-962
FAM-xtra CPG 1000 Å
FAM-xtra CPG
Fluorescein CPGs have traditionally been used to add the fluorescein label at the 3'-terminus.
  • Stable for 12 months
Quality tested in-house
Caution: For Research Use Only