CM-DiI, CytoTrace™
Supplier: AAT BIOQUEST INC
22057 22058
77461-902EA 241.28 CAD
77461-902 77461-904
CM-DiI, CytoTrace™
CM-DiI
1,1'-Dioctadecyl-3,3,3',3'-tetramethylindocarbocyanine iodide (CM-DiI) is a lipophilic carbocyanine dye that is commonly used for labeling cell membranes in various biological and neuroscientific research applications.
  • Stable for 12 months
Quality tested in-house
Caution: For Research Use Only