Calbryte™ 520 potassium salt
Supplier: AAT BIOQUEST INC
20657
77461-768EA
1191.16
CAD
77461-768
Calbryte™ 520 potassium salt
Calbryte™ 520 potassium salt
Calbryte™ 520 is a new generation of fluorescent indicator for calcium measurements; its greatly improved signal-to-noise ratio and better intracellular retention properties make Calbryte™ 520 the most robust calcium indicator.
- Stable for 12 months
Calcium measurement is critical for numerous biological investigations.
Caution: For Research Use Only