Bio-16-UTP [Biotin-16-UTP] 1 mM
Supplier: AAT Bioquest
17111
17110
77461-544EA
660.09
CAD
77461-544
77461-542
Bio-16-UTP [Biotin-16-UTP] 1 mM
Bio-16-UTP [Biotin-16-UTP]
Bio-16-UTP is biotinylated UTP that can be used as a substrate in in vitro transcription reactions with a variety of RNA polymerases, including T7 RNA polymerase, SP6 RNA polymerase, or T3 RNA polymerase.
- Stable for 12 months
Quality tested in-house
Caution: For Research Use Only