Amino-Modifier C6 dT
Supplier: AAT BIOQUEST INC
6002
77462-938EA
248.08
CAD
77462-938
Amino-Modifier C6 dT
Amino-Modifier C6 dT
Amino-Modifier C6 dT is a convenient phosphoramidite reagent used to introduce an amino group from dT in oligo synthesis.
- Stable for 6 months
Quality tested in-house
MW:
995.05 g/mol |
CAS Number:
178925-21-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...