Eppendorf® Xplorer® Electronic Single-Channel Pipettes
Supplier: Eppendorf
4861000015 4861000023 4861000040 4861000058 4861000066 4861000031 4861000017 4861000027 4861000044
CA89133-424EA 1394.15 CAD
CA89133-424 CA89133-426 CA89133-430 CA89133-432 CA89133-434 CA89133-428 CA76420-654 CA76420-656 CA76420-658
Eppendorf® Xplorer® Electronic Single-Channel Pipettes
Pipettes Micropipettes
Eppendorf Xplorer single-channel pipettes provide reliable, ergonomic high-precision liquid handling. With easy operation, five pipetting modes, and advanced charging technology, they minimize errors and boost reproducibility in a range of lab applications.
  • Electronic single channel pipettes designed to aspirate and dispense liquid volumes from 0.5 μl to 10 ml
  • Intuitive operating concept and ergonomic design
  • Multifunctional rocker with ergonomic display angle
  • Selection dial and innovative ejector
  • Convenient handle and powerful rechargeable battery
  • Flexible
More Product Information