UltraTemp® Ultra-Thin ESD Polyamide Circuit Board Labels for Printers
Supplier: Brady Worldwide
176949 176950 176951 176952 176953 176954 176955 176956 176957 176942 176943 176944
77574-862RL 269.41 CAD
77574-862 77574-906 77574-922 77574-860 77574-944 77574-870 77574-934 77574-898 77574-880 77574-912 77574-928 77574-876
UltraTemp® Ultra-Thin ESD Polyamide Circuit Board Labels for Printers
Labels
Ideal for PCB and electronic component manufacturing, these labels are static dissipative and survive extreme processes like wave soldering and multiple harsh washes.
  • Engineered to withstand high temperatures
  • Low profile, glossy white polyimide material (B-768) features
  • Static dissipative adhesive and liner
  • Labels do not require reflow or preheatings to ensure printing readability
  • Service temperature of −70 to +100 °C (−94 to +212 °F)
Enhanced topcoat is ideal for boards that go through multiple harsh-condition washes. A ribbon is required to print this material. Use Part Number M7-R6300 for the M710/BMP71 label printers. Surface resistivity meets ANSI/ESD S541-2008 recommended range for dissipative ESD packaging materials.
Certifications: UL approved; meets requirements of MIL-STD-202G, method 215K.