Eppendorf® Xplorer® Plus Move It® Electronic Multi-Channel Pipettes With Adjustable Tip Spacing
Supplier: Eppendorf
4861000816 4861000817 4861000818 4861000833 4861000834 4861000835 4861000816 4861000817 4861000818 4861000833 4861000834 4861000835 4861000781 4861000794 4861000782 4861000795 4861000781 4861000794 4861000782 4861000795
76452-838EA 5132.33 CAD
76452-838 76452-840 76452-842 76452-844 76452-846 76452-848 CA76474-592 CA76474-594 CA76474-596 CA76474-598 CA76474-600 CA76474-602 76541-126 76541-128 76541-130 76541-132 CA76541-126 CA76541-128 CA76541-130 CA76541-132
Eppendorf® Xplorer® Plus Move It® Electronic Multi-Channel Pipettes With Adjustable Tip Spacing
Pipettes Micropipettes
Eppendorf® Xplorer® Plus Move It® Electronic multi-channel pipettes with adjustable cones allow easy vessel format changes, cutting up to 50% of your time compared to single-channel pipettes. With several pipetting modes for enhanced productivity and offered in 4, 6, 8 or 12 channels.
  • Air-cushion pipettes with variable volume and adjustable tip spacing are available in 4, 6, 8 and 12-channel
  • Choose from Mechanical (Research™ plus) or Electronic (Xplorer™ plus) adjustable tip spacing multichannel pipettes
  • For Plates (12, 24, 48, 96, and 384-well), Tubes (1.5
More Product Information