Eppendorf Xplorer® Electronic Multi-Channel Pipettes
Supplier: Eppendorf
4861000112 4861000120 4861000139 4861000147 4861000155 4861000163 4861000104 4861000171 4861000171
CA89133-438EA 2873.95 CAD
CA89133-438 CA89133-440 CA89133-442 CA89133-444 CA89133-446 CA89133-448 CA89133-436 89496-610 CA89496-610
Eppendorf Xplorer® Electronic Multi-Channel Pipettes
Pipettes Micropipettes
Eppendorf Xplorer® multi-channel pipettes with dispensing mode provide a cutting-edge solution for high-throughput liquid handling. Offering the highest levels of simplicity, precision, and reproducibility, these pipettes will help you pipette with more speed, comfort, and ergonomics.
  • Electronic single, 8 and 12 channel pipettes designed to aspirate and dispense liquid volumes from 0.5 μl to 10 ml
  • Intuitive operating concept and ergonomic design
  • Multifunctional rocker and ergonomic display angle
  • Selection dial and innovative ejector
  • Convenient handle with
More Product Information