ReadiUse™ ddNTP Terminator Mix *10 mM*
Supplier: AAT BIOQUEST INC
17205
17206
77461-548EA
341.44
CAD
77461-548
77461-550
ReadiUse™ ddNTP Terminator Mix *10 mM*
Nucleic Acid Reagents
Nucleotides
ReadiUse™ ddNTP Terminator Mix consists of 4 equimolar 2',3'-ddNTPs, including ddATP, ddGTP, ddTTP and ddCTP.
- All the 2',3'-ddNTPs are highly purified
They are provided in 10 mM solution and can be readily added to a sequencing solution or other biological samples.
Caution: For research use only.