5-Azacytidine >98% (by TLC)
Supplier: Sino Biological
|
Danger
|
D353-904-250
D353-904-50
77556-262EA
200.66
CAD
77556-262
77556-264
5-Azacytidine >98% (by TLC)
5-Azacytidine
Formula:
C₈H₁₂N₄O₅ Melting Pt: 226...232 °C (dec.) Storage Temperature: Freezer |
MDL Number:
MFCD00006539 CAS Number: 320-67-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...