N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-(3-morpholinopropoxy)quinazolin-4-amine >98% (by TLC, NMR)

Supplier: Sino Biological
Warning

E10-900-1000 E10-900-200
77556-894EA 443.98 CAD
77556-894 77556-896
N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-(3-morpholinopropoxy)quinazolin-4-amine >98% (by TLC, NMR)
N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-(3-morpholinopropoxy)quinazolin-4-amine
Formula: C₂₂H₂₄ClFN₄O₃
Storage Temperature: Ambient
MDL Number: MFCD04307832
CAS Number: 184475-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR