CytoWatch™ Multiplexing Buffer for Flow Cytometry, 20X
Supplier: AAT BIOQUEST INC
37010 37011
77462-200EA 133.84 CAD
77462-200 77462-202
CytoWatch™ Multiplexing Buffer for Flow Cytometry, 20X
Buffers Cell Culture Buffers
CytoWatch™ Multiplexing buffer is a buffer optimized for the multicolor flow cytometric analysis of cells.
  • Designed for ease of use
  • Enhances multicolor flow experiments with two or more staining reagents
CytoWatch™ Multiplexing buffer is a concentrated solution that is directly added to the staining solutions of certain fluorescent antibody conjugates or other fluorescent reagents before staining cells.
Caution: For research use only.