CytoWatch™ Wash-free Fluorescence Cell Imaging Buffer, 10X
Supplier: AAT BIOQUEST INC
20045 20046
77461-732EA 159.49 CAD
77461-732 77461-734
CytoWatch™ Wash-free Fluorescence Cell Imaging Buffer, 10X
Protein Linking and Labeling Systems
CytoWatch™ Wash-free fluorescence cell imaging buffer *10X* is a ready-to-use buffer optimized for fluorescence cell imaging.
  • Designed for ease of use
  • Saves time
CytoWatch™ Wash-free fluorescence cell imaging buffer is 10X concentrated and should be diluted to 1X with PBS before use.
Caution: For research use only.