(R)-Roscovitine >99% (by TLC, NMR)
Supplier: Sino Biological
|
|
C22-901-02
C22-901-10
77555-226EA
197.5
CAD
77555-226
77555-228
(R)-Roscovitine >99% (by TLC, NMR)
(R)-Roscovitine
Formula:
C₁₉H₂₆N₆O Storage Temperature: Freezer |
MDL Number:
MFCD02266401 CAS Number: 186692-46-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...