NSC 663284 >98%

Supplier: Sino Biological

C04-902-25 C04-902-5
77554-962EA 644.64 CAD
77554-962 77554-964
NSC 663284 >98%
NSC 663284
Formula: C₁₅H₁₆ClN₃O₃
Storage Temperature: Freezer
MDL Number: MFCD08276924
CAS Number: 383907-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR