SC-79 98% (by TLC, NMR)

Supplier: Sino Biological

A16-901-05 A16-901-25
77554-104EA 110.6 CAD
77554-104 77554-106
SC-79 98% (by TLC, NMR)
SC-79
Formula: C₁₇H₁₇ClN₂O₅
Storage Temperature: Freezer
MDL Number: MFCD02681303
CAS Number: 305834-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR