Logic® Particle Counter Integration Kit

Supplier: Labconco
Logic® Mag-Zone™
3034000
77574-624EA 402.9 CAD
77574-624
Logic® Particle Counter Integration Kit
Particle Counters
Particle counter integration kit for Logic Biosafety cabinet. Compatible with 4 to 20 mA output.

  • For use with user supplied particle counter
  • Applicable to Logic Biosafety cabinets 2024 or newer models
  • 115 V, 15 A
  • Weight: 3.0 lbs.
  • Dimensions: 2W×29D×1" H
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR