Twist Lock Power Cord
Supplier: Labconco
1366800 1366900
77574-630EA 161.16 CAD
77574-630 77574-632
Twist Lock Power Cord
Cables
Twist Lock Power Cord, 15A - IEC-C13 connector with NEMA L5-15 plug.
Power your biosafety cabinet with Labconco's twist-lock power cord. This model is for use in North America at 115 V and features a 15A - IEC-C13 connector with a NEMA L5-15 plug. The electrical cord is a generous 12' in length.