PI(5)P:PS Lipid Substrates
Supplier: Sino Biological
P428-59-1000 P428-59-500
77561-816EA 357.08 CAD
77561-816 77561-818
PI(5)P:PS Lipid Substrates
Assays Protein Assays
The substrate solution used for assaying lipid kinases.
Formulation: Frozen substrate solution containing 0.25 mM of Phospha-tidylinositol 5-phosphate [PI(5)P or PtdIns5P] and 2 mM of Phosphatidylserine (PS) in 1X lipid dilution buffer
For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.