PI(3)P:PS Lipid Substrates
Supplier: Sino Biological
P426-59-1000 P426-59-500
77561-808EA 357.08 CAD
77561-808 77561-810
PI(3)P:PS Lipid Substrates
Assays Protein Assays
The substrate solution used for assaying lipid kinases.
Formulation: Frozen substrate solution containing 0.25 mM of Phospha-tidylinositol 3-phosphate [PI(3)P or PtdIns3P] and 2 mM of Phosphatidylserine (PS) in 1X lipid dilution buffer
For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.