PI(5)P Lipid Substrates
Supplier: Sino Biological
P423-59-1000 P423-59-500
77561-796EA 687.3 CAD
77561-796 77561-798
PI(5)P Lipid Substrates
Assays Protein Assays
The substrate solution used for assaying lipid kinases.
Formulation: Frozen substrate solution containing 1 mM of Phospha-tidylinositol 5-phosphate [PI(5)P or PtdIns5P] in 1X lipid dilution buffer
For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.