BradyJet™ J7300 Color Label Printer WiFi with Product and Wire ID Software
Supplier: Brady
BradyJet™ J7300
176879 176880
77580-942EA 3779.68 CAD
77580-942 77581-036
BradyJet™ J7300 Color Label Printer WiFi with Product and Wire ID Software
Printers Label Printers
Unleash your true colours with the BradyJet™ J7300 Colour Label Printer. Create vibrant, industrial-grade inkjet for safety labels, signs and tags in full colour. Non-WiFi option available.
  • Full color printing capability
  • PC connected
  • Print resolution: 4800 dpi
  • 2 year warranty
  • Maximum label width: 4,25"
  • Maximum labels per day: 5000
Ideal for industrial labeling that changes frequently, from UL product/component ID and instructional labels to work in process, asset, and bin labeling. LabelSense™ technology enables printer, materials, and inks to talk to each other for automatic part recognition, setup, and adjustments. Dual CMY and true black (K) ink cartridges deliver vibrant colours and deeper darks for customized colours, headers, and logos, and sharp barcodes and text. Prints smear-free inks on labels that resist water, chemicals, and abrasions for up to two-year outdoor durability (even longer indoors). Print Utility dashboard helps users plan, budget, and troubleshoot with status alerts, a job cost calculator, remote monitoring ,and more. Exterior LED lighting and clear cover communicate real-time printer status.
Brady Workstation Product and Wire ID Suite helps create labels with text, graphics, and custom formatting, or by serializing and importing data. Optimized Windows® driver allows connection to third-party software and print PDFs.