Chemical-Resistant Polypropylene Label Tapes for BradyJet™ J7300 Color Label Printer
Supplier: Brady
176608 176607 176617 176606 170478 176618 176609 176584 176631 170463 176636 176610 176634 176585 170479 176615 176616 176633 176635 176613 176614 176611 170464
77581-102RL 393.1 CAD
77581-102 77581-028 77581-006 77581-016 77581-078 77581-034 77581-056 77580-918 77580-964 77580-920 77580-930 77580-984 77580-994 77580-970 77580-992 77581-040 77581-094 77580-904 77580-936 77581-074 77581-080 77580-934 77580-988
Chemical-Resistant Polypropylene Label Tapes for BradyJet™ J7300 Color Label Printer
Labels
Create full-color identification and barcode labels that outlast liquid nitrogen, autoclave conditions, and harsh chemicals.
  • Permanent acrylic adhesive
  • Matte finish
  • Chemical-resistant material
  • Suitable for low temperature application
  • Service temperature range: –112 to +158 °F
  • RoHS compliant to 2011/65/EU and 2015/863
Inkjet receptive label material is ideal for full-colour laboratory identification on vials, centrifuge tubes, and test tubes. Labels withstand extreme temperatures from -112 to 158° F (-80 to 70° C). Permanent acrylic adhesive bonds tightly to smooth surfaces like glass, copper, and stainless steel. Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup, and seamless software interaction.
For use with the BradyJet™ J7300 colour Label Printer. Ink cartridges are required to print on this material (use part numbers P-CMY-1 and P-K-1).